Assay Details
Target Gene Details
Entrez Gene ID: | 7818 |
Gene Name: | death associated protein 3 |
Gene Aliases: |
DAP-3, MRP-S29, MRPS29, bMRP-10 |
Location: |
Chr.1:155688104-155739010 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAP3 | XM_017002291.1 | XP_016857780.1 | ||
| XM_017002292.1 | XP_016857781.1 | |||
| XM_017002294.1 | XP_016857783.1 | |||
| XM_017002295.1 | XP_016857784.1 | |||
| AK022879.1 | 1 | 652 |
Target Gene Details
Entrez Gene ID: | 55249 |
Gene Name: | YY1 associated protein 1 |
Gene Aliases: |
HCCA1, HCCA2, YAP, YY1AP |
Location: |
Chr.1:155659442-155689032 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YY1AP1 | NM_001198900.1 | NP_001185829.1 | ||
| NM_001198901.1 | NP_001185830.1 | |||
| NM_001198902.1 | NP_001185831.1 | |||
| NM_001198903.1 | NP_001185832.1 | |||
| NM_001198904.1 | NP_001185833.1 | |||
| NM_001198905.1 | NP_001185834.1 | |||
| NM_001198906.1 | NP_001185835.1 | |||
| NM_018253.3 | NP_060723.2 | |||
| NM_139118.2 | NP_620829.1 | |||
| NM_139119.2 | NP_620830.1 | |||
| NM_139121.2 | NP_620832.1 | |||
| AF206328.1 | AAL74055.1 | |||
| AF466401.1 | AAL75971.1 | |||
| AK023976.1 | BAB14748.1 | |||
| AK298904.1 | ||||
| AK315049.1 | ||||
| AY604179.1 | AAT34990.1 | |||
| BC001655.2 | AAH01655.2 | |||
| BC001843.2 | AAH01843.2 | |||
| BC014906.2 | AAH14906.1 | |||
| BC044887.1 | AAH44887.1 | |||
| BG325066.1 | ||||
| CR749789.1 | CAH18650.1 | |||
| DC300265.1 | ||||
| DC388975.1 | ||||
| DC399389.1 | ||||
| DC406177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436522 | Chr.1:155589722 - 155704458 on Build GRCh38 | Deletion |
|
| esv3578199 | Chr.1:155687166 - 155693640 on Build GRCh38 | Loss |
|
| nsv946421 | Chr.1:155686439 - 155689285 on Build GRCh38 | Duplication |
|
| esv33869 | Chr.1:155253492 - 155948170 on Build GRCh38 | Loss |
|
| nsv428246 | Chr.1:155588406 - 155816040 on Build GRCh38 | Gain |
|
| esv3587627 | Chr.1:155619701 - 155695480 on Build GRCh38 | Loss |
|
| esv2718429 | Chr.1:155648347 - 155763436 on Build GRCh38 | Deletion |
|
| nsv3054 | Chr.1:155684044 - 155703718 on Build GRCh38 | Deletion |
|
| nsv547965 | Chr.1:155593361 - 155718809 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map