Assay Details
Target Gene Details
Entrez Gene ID: | 280664 |
Gene Name: | WAP four-disulfide core domain 10B |
Gene Aliases: |
WAP12 |
Location: |
Chr.20:45684651-45708899 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC10B | XM_017027824.1 | XP_016883313.1 | ||
| XM_017027825.1 | XP_016883314.1 |
Target Gene Details
Entrez Gene ID: | 164237 |
Gene Name: | WAP four-disulfide core domain 13 |
Gene Aliases: |
C20orf138, WAP13, dJ601O1.3 |
Location: |
Chr.20:45702016-45708817 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC13 | NM_172005.1 | 4 | 724 | NP_742002.1 |
| AF454505.1 | 4 | 724 | AAN70988.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv586081 | Chr.20:45573240 - 45747557 on Build GRCh38 | Loss |
|
| esv2751915 | Chr.20:45708077 - 45749572 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map