Assay Details
Target Gene Details
Entrez Gene ID: | 9129 |
Gene Name: | pre-mRNA processing factor 3 |
Gene Aliases: |
HPRP3, HPRP3P, PRP3, Prp3p, RP18, SNRNP90 |
Location: |
Chr.1:150321479-150353233 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRPF3 | NM_004698.2 | NP_004689.1 | ||
XM_011510129.1 | XP_011508431.1 | |||
XM_011510131.2 | XP_011508433.1 | |||
XM_011510132.2 | XP_011508434.1 | |||
XM_017002791.1 | XP_016858280.1 | |||
AF001947.1 | AAC09069.1 | |||
AF016370.1 | AAC51926.1 | |||
AK222825.1 | BAD96545.1 | |||
AK297139.1 | ||||
AK297265.1 | ||||
AK299980.1 | ||||
AK309330.1 | ||||
AK310241.1 | ||||
AK311653.1 | ||||
AK312891.1 | ||||
BC000184.2 | AAH00184.1 | |||
BC001954.1 | AAH01954.1 | |||
DC407158.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831548 | Chr.1:150296285 - 150463240 on Build GRCh38 | Loss |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -




Gene Ontology Categories:
