Assay Details
Target Gene Details
Entrez Gene ID: | 64207 |
Gene Name: | interferon regulatory factor 2 binding protein like |
Gene Aliases: |
C14orf4, EAP1 |
Location: |
Chr.14:77024543-77028699 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IRF2BPL | NM_024496.3 | 1 | 1677 | NP_078772.1 |
| AB058768.1 | 2 | 1178 | BAB47494.1 | |
| AW001869.1 |
More Information
Set Membership: |
|

Genomic Map