Assay Details
Target Gene Details
Entrez Gene ID: | 79712 |
Gene Name: | glycosyltransferase like domain containing 1 |
Gene Aliases: |
Hmat-Xa, mat-Xa |
Location: |
Chr.2:143945787-144333092 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 27 |
Target Gene Details
Entrez Gene ID: | 101928386 |
Gene Name: | uncharacterized LOC101928386 |
Gene Aliases: |
- |
Location: |
Chr.2:143937067-143964156 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928386 | NR_110237.1 |
More Information
Set Membership: |
|

Genomic Map