Assay Details
Target Gene Details
Entrez Gene ID: | 3091 |
Gene Name: | hypoxia inducible factor 1 alpha subunit |
Gene Aliases: |
HIF-1-alpha, HIF-1A, HIF-1alpha, HIF1, HIF1-ALPHA, MOP1, PASD8, bHLHe78 |
Location: |
Chr.14:61695401-61748259 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIF1A | NM_001243084.1 | 15 | 3565 | NP_001230013.1 |
| NM_001530.3 | 15 | 3668 | NP_001521.1 | |
| NM_181054.2 | 14 | 3541 | NP_851397.1 | |
| AB073325.1 | 14 | 3165 | BAB70608.1 | |
| AJ227916.1 | 1 | 80 | ||
| BC012527.2 | 15 | 3518 | AAH12527.1 | |
| BX648795.1 | 15 | 3532 | ||
| U22431.1 | 15 | 3292 | AAC50152.1 | |
| U29165.1 | 15 | 3528 | AAC51210.1 | |
| X72726.1 | 1 | 866 |
Target Gene Details
Entrez Gene ID: | 100750247 |
Gene Name: | HIF1A antisense RNA 2 |
Gene Aliases: |
3'aHIF-1A, aHIF |
Location: |
Chr.14:61747039-61749089 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIF1A-AS2 | NR_045406.1 | 1 | 1222 | |
| U85044.1 | 1 | 1082 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1300 | Chr.14:61737786 - 61782490 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map