Assay Details
Target Gene Details
Entrez Gene ID: | 94101 |
Gene Name: | ORMDL sphingolipid biosynthesis regulator 1 |
Gene Aliases: |
- |
Location: |
Chr.2:189766149-189784371 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORMDL1 | NM_001128150.1 | 2 | 166 | NP_001121622.1 |
| NM_016467.4 | 3 | 276 | NP_057551.1 | |
| XM_005246970.2 | 2 | 195 | XP_005247027.1 | |
| XM_011512199.2 | 2 | 158 | XP_011510501.1 | |
| XM_011512200.2 | 1 | 1773 | XP_011510502.1 | |
| XM_017005347.1 | 3 | 315 | XP_016860836.1 | |
| XM_017005348.1 | 2 | 147 | XP_016860837.1 | |
| AB064959.1 | 1 | 735 | BAC11710.1 | |
| AB064960.1 | 2 | 174 | BAC11711.1 | |
| AF151036.1 | 2 | 115 | AAF36122.1 | |
| AF395704.1 | 3 | 231 | AAM43503.1 | |
| AK074756.1 | 2 | 165 | BAC11184.1 | |
| AK075160.1 | 1 | 167 | BAC11441.1 | |
| AK313533.1 | 3 | 241 | ||
| BC005200.1 | 2 | 133 | AAH05200.1 | |
| CR936635.1 | 1 | 843 | CAI56776.1 | |
| DB122266.1 | 2 | 166 |
More Information
Set Membership: |
|

Genomic Map