Assay Details
Target Gene Details
Entrez Gene ID: | 26168 |
Gene Name: | SUMO1/sentrin/SMT3 specific peptidase 3 |
Gene Aliases: |
SMT3IP1, SSP3, Ulp1 |
Location: |
Chr.17:7561992-7571969 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP3 | NM_015670.5 | 2 | 458 | NP_056485.2 |
| AF199459.1 | 2 | 245 | AAL25652.1 | |
| AL557688.3 | 2 | 497 | ||
| AL834294.1 | 2 | 252 | CAD38967.1 | |
| AY008763.1 | 2 | 226 | AAG33252.1 | |
| BC080658.1 | 2 | 440 | AAH80658.1 | |
| BQ956948.1 | 2 | 458 | ||
| CX165088.1 | 2 | 394 |
Target Gene Details
Entrez Gene ID: | 100533955 |
Gene Name: | SENP3-EIF4A1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.17:7561992-7579006 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP3-EIF4A1 | NR_037926.1 | 2 | 458 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map