Assay Details
Target Gene Details
Entrez Gene ID: | 64218 |
Gene Name: | semaphorin 4A |
Gene Aliases: |
CORD10, RP35, SEMAB, SEMB |
Location: |
Chr.1:156149673-156177751 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA4A | NM_001193300.1 | 12 | 1618 | NP_001180229.1 |
| NM_001193301.1 | 12 | 1543 | NP_001180230.1 | |
| NM_001193302.1 | 10 | 1313 | NP_001180231.1 | |
| NM_022367.3 | 12 | 1627 | NP_071762.2 | |
| XM_011509871.2 | 11 | 1321 | XP_011508173.1 | |
| XM_011509872.2 | 13 | 1671 | XP_011508174.1 | |
| XM_011509873.2 | 13 | 1662 | XP_011508175.1 | |
| XM_011509874.2 | 11 | 1405 | XP_011508176.1 | |
| XM_011509875.2 | 11 | 1460 | XP_011508177.1 | |
| XM_011509876.2 | 11 | 1370 | XP_011508178.1 | |
| XM_011509878.2 | 11 | 1329 | XP_011508180.1 | |
| XM_011509879.2 | 11 | 1317 | XP_011508181.1 | |
| XM_017002056.1 | 11 | 1395 | XP_016857545.1 | |
| XM_017002057.1 | 13 | 1689 | XP_016857546.1 | |
| AB029394.1 | 11 | 1575 | BAB20087.1 | |
| AK022349.1 | 2 | 170 | BAB14018.1 | |
| AK091127.1 | 10 | 1313 | ||
| AK290353.1 | 12 | 1442 | ||
| AK296693.1 | 12 | 1336 | ||
| AK315547.1 | 12 | 1564 | ||
| AY358531.1 | 12 | 1468 | AAQ88895.1 | |
| BC020974.1 | 12 | 1561 | AAH20974.1 | |
| BX640891.1 | 13 | 1466 | CAE45942.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509513 | Chr.1:155932322 - 156178790 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map