Assay Details
Target Gene Details
Entrez Gene ID: | 5799 |
Gene Name: | protein tyrosine phosphatase, receptor type N2 |
Gene Aliases: |
IA-2beta, IAR, ICAAR, PTPRP, R-PTP-N2 |
Location: |
Chr.7:157539052-158587802 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRN2 | NM_001308267.1 | NP_001295196.1 | ||
| NM_001308268.1 | NP_001295197.1 | |||
| NM_002847.4 | NP_002838.2 | |||
| NM_130842.3 | NP_570857.2 | |||
| NM_130843.3 | NP_570858.2 | |||
| XM_017012475.1 | XP_016867964.1 | |||
| AB002385.2 | ||||
| AL157451.1 | CAB75665.2 | |||
| AW292950.1 | ||||
| BC034040.1 | AAH34040.1 | |||
| BG059906.1 | ||||
| CB850986.1 | ||||
| U66702.1 | AAC50742.1 | |||
| U81561.1 | AAB68603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3615699 | Chr.7:157524033 - 157541939 on Build GRCh38 | Gain |
|
| nsv8242 | Chr.7:157493009 - 157555855 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map