Assay Details
Target Gene Details
Entrez Gene ID: | 5303 |
Gene Name: | peptidylprolyl cis/trans isomerase, NIMA-interacting 4 |
Gene Aliases: |
EPVH, PAR14, PAR17 |
Location: |
Chr.X:72181676-72263964 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIN4 | NM_001170747.1 | NP_001164218.1 | ||
| NM_006223.3 | 4 | 946 | NP_006214.2 | |
| NR_033187.1 | 3 | 872 | ||
| AB009690.1 | 4 | 867 | BAA82320.1 | |
| AF143096.1 | 4 | 860 | AAD27893.1 | |
| AK127605.1 | ||||
| BC005234.1 | 4 | 865 | AAH05234.2 | |
| BC070288.1 | 4 | 865 | AAH70288.1 | |
| BC111394.1 | 4 | 869 | ||
| BE504461.1 | 1 | 126 | ||
| BU952002.1 | 1 | 386 |
More Information
Set Membership: |
|

Genomic Map