Assay Details
Target Gene Details
Entrez Gene ID: | 6462 |
Gene Name: | sex hormone binding globulin |
Gene Aliases: |
ABP, SBP, TEBG |
Location: |
Chr.17:7614064-7633383 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHBG | NM_001040.4 | 6 | 854 | NP_001031.2 |
| NM_001146279.2 | 6 | 800 | NP_001139751.1 | |
| NM_001146280.2 | 6 | 854 | NP_001139752.1 | |
| NM_001146281.2 | NP_001139753.1 | |||
| NM_001289113.1 | 6 | 847 | NP_001276042.1 | |
| NM_001289114.1 | 6 | 710 | NP_001276043.1 | |
| NM_001289115.1 | 6 | 847 | NP_001276044.1 | |
| NM_001289116.1 | 6 | 762 | NP_001276045.1 | |
| AK302603.1 | 6 | 847 | ||
| BC069597.1 | 6 | 772 | AAH69597.1 | |
| BC101785.1 | 6 | 806 | AAI01786.1 | |
| BC112186.1 | 6 | 806 | AAI12187.1 | |
| CD013955.1 | 6 | 796 | ||
| CD013957.1 | 6 | 742 | ||
| CD013958.1 | 6 | 704 | ||
| EU352661.1 | ||||
| EU352662.1 | 5 | 532 | ||
| EU352663.1 | ||||
| EU352664.1 | ||||
| EU352665.1 | 6 | 715 | ||
| EU352666.1 | ||||
| EU352667.1 | 5 | 553 | ||
| EU352668.1 | ||||
| EU352669.1 | ||||
| EU352670.1 | 6 | 710 | ||
| EU352671.1 | 6 | 710 | ||
| EU352672.1 | ||||
| EU352673.1 | ||||
| EU352674.1 | ||||
| EU352675.1 | 5 | 548 | ||
| EU352676.1 | 5 | 548 | ||
| X05403.1 | 6 | 712 | CAA28987.1 | |
| X05792.1 | 4 | 417 | CAA29234.1 | |
| X05885.1 | 5 | 639 | CAA29309.1 | |
| X16351.1 | 6 | 760 | CAA34400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map