Assay Details
Target Gene Details
Entrez Gene ID: | 10235 |
Gene Name: | RAS guanyl releasing protein 2 |
Gene Aliases: |
CALDAG-GEFI, CDC25L |
Location: |
Chr.11:64726911-64745456 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv818842 | Chr.11:64577976 - 64778919 on Build GRCh38 | Gain |
|
| nsv359 | Chr.11:64709536 - 64735401 on Build GRCh38 | Deletion |
|
| dgv1965n54 | Chr.11:64566642 - 64806117 on Build GRCh38 | Loss |
|
| dgv1967n54 | Chr.11:64709578 - 64817487 on Build GRCh38 | Loss |
|
| nsv832189 | Chr.11:64719711 - 64852727 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map