Assay Details
Target Gene Details
Entrez Gene ID: | 89876 |
Gene Name: | MYCBP associated and testis expressed 1 |
Gene Aliases: |
AAT1, AAT1alpha, C3orf15, CaM-IP2, SPATA26 |
Location: |
Chr.3:119703022-119767102 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAATS1 | NM_001320316.1 | NP_001307245.1 | ||
| NM_001320317.1 | NP_001307246.1 | |||
| NM_001320318.1 | NP_001307247.1 | |||
| NM_033364.3 | NP_203528.2 | |||
| XM_011513277.1 | XP_011511579.1 | |||
| AF440403.1 | AAP97317.1 | |||
| AF497717.1 | AAM19218.1 | |||
| AK055558.1 | ||||
| AK289914.1 | ||||
| AK294413.1 | ||||
| AK302699.1 | ||||
| AK303997.1 | ||||
| AK309317.1 | ||||
| AL133066.1 | CAB61389.1 | |||
| BC035248.1 | AAH35248.1 | |||
| BC126394.1 | ||||
| CR749242.1 | CAH18098.1 | |||
| DB445414.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460834 | Chr.3:119665316 - 119740004 on Build GRCh38 | Gain |
|
| nsv591391 | Chr.3:119684298 - 119723725 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114121988] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map