Assay Details
Target Gene Details
Entrez Gene ID: | 22883 |
Gene Name: | calsyntenin 1 |
Gene Aliases: |
ALC-ALPHA, CDHR12, CST-1, CSTN1, PIK3CD, XB31alpha, alcalpha1, alcalpha2 |
Location: |
Chr.1:9729021-9824526 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLSTN1 | NM_001009566.2 | 19 | 4291 | NP_001009566.1 |
| NM_001302883.1 | 18 | 4234 | NP_001289812.1 | |
| NM_014944.4 | 18 | 4261 | NP_055759.3 | |
| AB020718.1 | 19 | 4291 | ||
| AF438482.1 | AAQ04552.1 | |||
| AK000605.1 | 14 | 2901 | ||
| AK001591.1 | 6 | 1569 | ||
| AK056820.1 | 4 | 1241 | ||
| AK225500.1 | 3 | 1019 | ||
| BC033902.2 | 18 | 3719 | AAH33902.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832203 | Chr.1:9666574 - 9800151 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map