Assay Details
Target Gene Details
Entrez Gene ID: | 154467 |
Gene Name: | coiled-coil domain containing 167 |
Gene Aliases: |
C6orf129, HSPC265 |
Location: |
Chr.6:37482920-37499924 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC167 | NM_138493.2 | 4 | 559 | NP_612502.1 |
| AF161383.1 | AAF28943.1 | |||
| BC003515.1 | AAH03515.1 | |||
| BC108655.1 | AAI08656.1 | |||
| BE907609.1 | 3 | 444 | ||
| DA152306.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752124 | Chr.6:37448443 - 37712846 on Build GRCh38 | Gain |
|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|

Genomic Map