Assay Details
Target Gene Details
Entrez Gene ID: | 3730 |
Gene Name: | anosmin 1 |
Gene Aliases: |
ADMLX, HH1, HHA, KAL, KAL1, KALIG-1, KMS, WFDC19 |
Location: |
Chr.X:8528874-8732187 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANOS1 | NM_000216.3 | 13 | 2076 | NP_000207.2 |
| BC137426.1 | 13 | 2040 | ||
| BC137427.1 | 13 | 2040 | ||
| CD642906.1 | ||||
| CN417843.1 | 3 | 393 | ||
| M97252.1 | 13 | 2075 | AAA59202.1 | |
| X60299.1 | 13 | 1988 | CAA42841.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv991240 | Chr.X:8489564 - 8545448 on Build GRCh38 | Gain |
|
| esv28971 | Chr.X:8485960 - 8549984 on Build GRCh38 | Gain |
|
| nsv518704 | Chr.X:8469952 - 8714909 on Build GRCh38 | Gain |
|
| esv2752339 | Chr.X:8398685 - 8910093 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map