Assay Details
Target Gene Details
Entrez Gene ID: | 5024 |
Gene Name: | purinergic receptor P2X 3 |
Gene Aliases: |
P2X3 |
Location: |
Chr.11:57335943-57372399 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RX3 | NM_002559.3 | 2 | 348 | NP_002550.2 |
| XM_011545069.2 | 3 | 1009 | XP_011543371.1 | |
| XM_011545070.2 | 3 | 315 | XP_011543372.1 | |
| XM_011545071.2 | 2 | 883 | XP_011543373.1 | |
| XM_011545072.2 | 3 | 1141 | XP_011543374.1 | |
| AB016608.1 | 2 | 164 | BAA76515.1 | |
| BC069162.1 | 2 | 240 | AAH69162.1 | |
| BC074792.2 | 2 | 195 | AAH74792.1 | |
| BC074793.2 | 2 | 195 | AAH74793.1 | |
| BC109287.2 | 2 | 198 | AAI09288.1 | |
| DB462670.1 | 2 | 348 |
More Information
Set Membership: |
|

Genomic Map