Assay Details
Target Gene Details
Entrez Gene ID: | 81794 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 10 |
Gene Aliases: |
ADAM-TS10, ADAMTS-10, WMS, WMS1 |
Location: |
Chr.19:8580240-8610735 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS10 | NM_001282352.1 | 13 | 1937 | NP_001269281.1 |
| NM_030957.3 | 26 | 3748 | NP_112219.3 | |
| XM_006722917.3 | 24 | 3741 | XP_006722980.1 | |
| XM_017027338.1 | 25 | 3604 | XP_016882827.1 | |
| XM_017027339.1 | 16 | 2228 | XP_016882828.1 | |
| XM_017027340.1 | 15 | 2088 | XP_016882829.1 | |
| AF163762.1 | AAG35563.1 | |||
| AK131414.1 | 13 | 1937 | BAD18561.1 | |
| BF588726.1 | 1 | 254 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv137n111 | Chr.19:8488438 - 8590905 on Build GRCh38 | Deletion |
|
| esv3643619 | Chr.19:8567215 - 8586980 on Build GRCh38 | Gain |
|
| esv2762024 | Chr.19:8567703 - 9197018 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map