Assay Details
Target Gene Details
Entrez Gene ID: | 127943 |
Gene Name: | Fc receptor like B |
Gene Aliases: |
FCRL2, FCRLM2, FCRLY, FREB-2, FcRY |
Location: |
Chr.1:161721544-161728143 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCRLB | NM_001002901.3 | 8 | 1825 | NP_001002901.1 |
| NM_001288829.1 | 6 | 1466 | NP_001275758.1 | |
| NM_001288830.1 | 5 | 1445 | NP_001275759.1 | |
| NM_001288831.1 | 6 | 1507 | NP_001275760.1 | |
| NM_001288832.1 | 5 | 1486 | NP_001275761.1 | |
| NM_001320241.1 | 7 | 1639 | NP_001307170.1 | |
| AK055614.1 | 2 | 1819 | BAB70971.1 | |
| AK290733.1 | 7 | 1639 | ||
| BC038564.2 | 3 | 1191 | AAH38564.2 | |
| BQ014014.1 | 1 | 189 | ||
| DQ021957.1 | 8 | 1825 | AAY42534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428257 | Chr.1:161436525 - 161898473 on Build GRCh38 | Loss |
|
| nsv464184 | Chr.1:161677017 - 161915562 on Build GRCh38 | Gain |
|
| nsv516773 | Chr.1:161721643 - 161732973 on Build GRCh38 | Gain |
|
| esv33393 | Chr.1:161508891 - 162110537 on Build GRCh38 | Gain+Loss |
|
| nsv464195 | Chr.1:161723213 - 161733002 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map