Assay Details
Target Gene Details
Entrez Gene ID: | 4170 |
Gene Name: | BCL2 family apoptosis regulator |
Gene Aliases: |
BCL2L3, EAT, MCL1-ES, MCL1L, MCL1S, Mcl-1, TM, bcl2-L-3, mcl1/EAT |
Location: |
Chr.1:150574551-150579738 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCL1 | NM_001197320.1 | 4 | 2780 | NP_001184249.1 |
| NM_021960.4 | 3 | 3239 | NP_068779.1 | |
| NM_182763.2 | 2 | 2991 | NP_877495.1 | |
| BC017197.2 | 3 | 3085 | AAH17197.1 | |
| L08246.1 | 3 | 3091 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006838 | Chr.1:150453465 - 150750307 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map