Assay Details
Target Gene Details
Entrez Gene ID: | 158787 |
Gene Name: | RIB43A domain with coiled-coils 1 |
Gene Aliases: |
2610028I09Rik |
Location: |
Chr.X:53422782-53431120 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RIBC1 | NM_001031745.4 | NP_001026915.1 | ||
| NM_001267053.3 | NP_001253982.1 | |||
| NM_144968.3 | 5 | 863 | NP_659405.1 | |
| XM_005261988.3 | XP_005262045.1 | |||
| XM_005261990.3 | XP_005262047.1 | |||
| AK057345.1 | 5 | 829 | BAB71438.1 | |
| AK290346.1 | 5 | 813 | ||
| AK292871.1 | ||||
| AK304179.1 | ||||
| BC036767.1 | AAH36767.1 | |||
| CF272701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
| nsv6908 | Chr.X:53412544 - 53446505 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map