Assay Details
Target Gene Details
Entrez Gene ID: | 84271 |
Gene Name: | DNA polymerase delta interacting protein 3 |
Gene Aliases: |
PDIP46, SKAR |
Location: |
Chr.22:42583721-42614962 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLDIP3 | NM_001278657.1 | 9 | 2225 | NP_001265586.1 |
| NM_032311.4 | 9 | 2174 | NP_115687.2 | |
| NM_178136.2 | 8 | 2087 | NP_835237.1 | |
| NR_103820.1 | 9 | 2163 | ||
| XM_011530457.1 | 8 | 2090 | XP_011528759.1 | |
| XM_017029010.1 | 7 | 1969 | XP_016884499.1 | |
| AB055760.2 | 9 | 2079 | ||
| AK024626.1 | 9 | 2103 | ||
| AL137743.1 | 9 | 2070 | CAH56378.1 | |
| AL160111.1 | 9 | 2092 | CAB77058.1 | |
| AL160112.1 | 8 | 2005 | CAB77059.1 | |
| AY506562.1 | AAR91612.1 | |||
| BC001488.2 | 9 | 2059 | AAH01488.2 | |
| BC019643.1 | 5 | 741 | AAH19643.1 | |
| BC049840.1 | 9 | 2081 | AAH49840.1 | |
| BC056912.1 | 8 | 1993 | AAH56912.2 | |
| BC095411.1 | 9 | 2151 | AAH95411.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3568423 | Chr.22:42522721 - 42586566 on Build GRCh38 | Loss |
|
| esv2758839 | Chr.22:42371195 - 42687300 on Build GRCh38 | Gain+Loss |
|
| nsv428391 | Chr.22:42371295 - 42698202 on Build GRCh38 | Gain |
|
| esv28197 | Chr.22:42483195 - 42609208 on Build GRCh38 | Gain |
|
| dgv4578n100 | Chr.22:42488885 - 42585161 on Build GRCh38 | Gain |
|
| dgv1323e212 | Chr.22:42559611 - 42586566 on Build GRCh38 | Loss |
|
| nsv9912 | Chr.22:42487331 - 42585567 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map