Assay Details
Target Gene Details
Entrez Gene ID: | 84932 |
Gene Name: | RAB2B, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.14:21459020-21476973 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB2B | NM_001163380.1 | 1 | 33 | NP_001156852.1 |
| NM_032846.3 | NP_116235.2 | |||
| NR_028074.1 | ||||
| XM_017021710.1 | XP_016877199.1 | |||
| XM_017021711.1 | XP_016877200.1 | |||
| AF468652.1 | AAN86142.1 | |||
| AK027730.1 | BAB55326.1 | |||
| AK296057.1 | ||||
| AK300599.1 | ||||
| AK307349.1 | ||||
| AK315354.1 | ||||
| BC020839.1 | AAH20839.1 | |||
| BC066366.1 | AAH66366.1 | |||
| BG535306.1 | 1 | 34 | ||
| BX647601.1 | CAI46103.1 | |||
| CD634472.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422459 | Chr.14:21463465 - 21701280 on Build GRCh38 | Duplication |
|
| esv3633716 | Chr.14:21455908 - 21564274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map