Assay Details
Target Gene Details
Entrez Gene ID: | 8836 |
Gene Name: | gamma-glutamyl hydrolase |
Gene Aliases: |
GH |
Location: |
Chr.8:63015079-63039051 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GGH | NM_003878.2 | NP_003869.1 | ||
| XM_011517623.2 | XP_011515925.1 | |||
| AK290450.1 | ||||
| AK301094.1 | ||||
| BC025025.1 | AAH25025.1 | |||
| U55206.1 | AAC05579.1 |
More Information
Set Membership: |
|

Genomic Map