Assay Details
Target Gene Details
Entrez Gene ID: | 94160 |
Gene Name: | ATP binding cassette subfamily C member 12 |
Gene Aliases: |
MRP9 |
Location: |
Chr.16:48082973-48146770 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCC12 | NM_033226.2 | NP_150229.2 | ||
| AF395908.1 | AAL79528.1 | |||
| AF395909.1 | AAL79529.1 | |||
| AF411577.1 | AAL99900.1 | |||
| AF411578.1 | AAL99901.1 | |||
| AY040220.1 | AAK76740.1 | |||
| AY196326.1 | AAO40749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520355 | Chr.16:47390917 - 48130866 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map