Assay Details
Target Gene Details
Entrez Gene ID: | 754 |
Gene Name: | pituitary tumor-transforming 1 interacting protein |
Gene Aliases: |
C21orf1, C21orf3, PBF |
Location: |
Chr.21:44849585-44873903 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTTG1IP | NM_001286822.1 | NP_001273751.1 | ||
| NM_004339.3 | NP_004330.1 | |||
| NR_104597.1 | ||||
| AF149785.1 | AAF73770.1 | |||
| AK094882.1 | ||||
| AK290139.1 | ||||
| AK293318.1 | ||||
| AK298557.1 | ||||
| AK299204.1 | ||||
| AK315626.1 | ||||
| BC000415.2 | AAH00415.1 | |||
| BC012858.1 | AAH12858.1 | |||
| BC019295.1 | AAH19295.1 | |||
| BC020983.1 | AAH20983.1 | |||
| BC031097.1 | AAH31097.1 | |||
| BC034250.1 | AAH34250.1 | |||
| BG721942.1 | ||||
| CN295711.1 | ||||
| L48984.1 | ||||
| Z50022.1 | CAA90325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058550 | Chr.21:44807248 - 44898019 on Build GRCh38 | Loss |
|
| esv3647136 | Chr.21:44802580 - 44905193 on Build GRCh38 | Loss |
|
| nsv459298 | Chr.21:44771532 - 44858053 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map