Assay Details
Target Gene Details
Entrez Gene ID: | 139170 |
Gene Name: | DDB1 and CUL4 associated factor 12 like 1 |
Gene Aliases: |
KIAA1892L, WDR40B |
Location: |
Chr.X:126549383-126552859 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF12L1 | NM_178470.4 | 2 | 3233 | NP_848565.2 |
| AI139786.1 | 1 | 171 | ||
| BC035674.1 | 2 | 3225 | AAH35674.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7078 | Chr.X:126537222 - 126569770 on Build GRCh38 | Insertion |
|
| nsv483075 | Chr.X:126448508 - 126598884 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map