Assay Details
Target Gene Details
Entrez Gene ID: | 1048 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 5 |
Gene Aliases: |
CD66e, CEA |
Location: |
Chr.19:41708585-41730433 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM5 | NM_001291484.2 | 8 | 2107 | NP_001278413.1 |
| NM_001308398.1 | 8 | 2104 | NP_001295327.1 | |
| NM_004363.5 | 8 | 2107 | NP_004354.3 | |
| XM_005258413.3 | XP_005258470.2 | |||
| XM_011526322.1 | 6 | 1542 | XP_011524624.1 | |
| XM_017026145.1 | 8 | 2073 | XP_016881634.1 | |
| XM_017026146.1 | 7 | 1821 | XP_016881635.1 | |
| AB852566.1 | 6 | 1426 | ||
| AB852567.1 | 5 | 892 | ||
| AK223101.1 | 8 | 2043 | BAD96821.1 | |
| BC034671.1 | 8 | 2055 | AAH34671.1 | |
| CR749337.1 | 6 | 1149 | CAH18191.1 | |
| M15042.1 | 8 | 1949 | AAA51963.1 | |
| M16234.1 | 4 | 971 | AAA51972.1 | |
| M29540.1 | 8 | 2074 | AAA51967.1 | |
| X16455.1 | 8 | 1946 | CAA34474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061309 | Chr.19:41683785 - 41724525 on Build GRCh38 | Gain |
|
| nsv482220 | Chr.19:41708611 - 41730516 on Build GRCh38 | Gain |
|
| nsv579615 | Chr.19:41717515 - 41806347 on Build GRCh38 | Gain |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map