Assay Details
Target Gene Details
Entrez Gene ID: | 139322 |
Gene Name: | apolipoprotein O like |
Gene Aliases: |
CXorf33, FAM121A, Mic27, UNQ8193 |
Location: |
Chr.X:85003852-85093317 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOOL | NM_198450.5 | 9 | 1567 | NP_940852.3 |
| AK130506.1 | 10 | 1539 | ||
| CR749250.1 | 10 | 1466 | CAH18106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740268 | Chr.X:85087907 - 85088704 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map