Assay Details
Target Gene Details
Entrez Gene ID: | 9152 |
Gene Name: | solute carrier family 6 member 5 |
Gene Aliases: |
GLYT-2, GLYT2, HKPX3, NET1 |
Location: |
Chr.11:20599400-20659285 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A5 | NM_001318369.1 | NP_001305298.1 | ||
| NM_004211.4 | NP_004202.3 | |||
| XM_017018545.1 | XP_016874034.1 | |||
| AF085412.1 | AAC95145.1 | |||
| BC096319.3 | AAH96319.1 | |||
| BC096320.3 | AAH96320.1 | |||
| BC096321.1 | AAH96321.1 | |||
| BC096322.3 | AAH96322.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509395 | Chr.11:20579204 - 20666939 on Build GRCh38 | Insertion |
|
| esv3891940 | Chr.11:20540113 - 20628616 on Build GRCh38 | Loss |
|
| nsv553678 | Chr.11:20097752 - 20718575 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map