Assay Details
Target Gene Details
Entrez Gene ID: | 84824 |
Gene Name: | Fc receptor like A |
Gene Aliases: |
FCRL, FCRL1, FCRLM1, FCRLX, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRX, FREB |
Location: |
Chr.1:161706972-161714352 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCRLA | NM_001184866.1 | 1 | 90 | NP_001171795.1 |
| NM_001184867.1 | 1 | 90 | NP_001171796.1 | |
| NM_001184870.1 | 1 | 90 | NP_001171799.1 | |
| NM_001184871.1 | 1 | 90 | NP_001171800.1 | |
| NM_001184872.1 | 1 | 90 | NP_001171801.1 | |
| NM_001184873.1 | 1 | 90 | NP_001171802.1 | |
| NM_032738.3 | 1 | 90 | NP_116127.3 | |
| XM_006711581.3 | XP_006711644.1 | |||
| XM_011510066.1 | XP_011508368.1 | |||
| AF531423.1 | 1 | 90 | AAM97591.1 | |
| BM471887.1 | 1 | 92 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428257 | Chr.1:161436525 - 161898473 on Build GRCh38 | Loss |
|
| nsv464184 | Chr.1:161677017 - 161915562 on Build GRCh38 | Gain |
|
| esv2758972 | Chr.1:161374465 - 161711762 on Build GRCh38 | Gain+Loss |
|
| esv33393 | Chr.1:161508891 - 162110537 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map