Assay Details
Target Gene Details
Entrez Gene ID: | 85313 |
Gene Name: | peptidylprolyl isomerase like 4 |
Gene Aliases: |
HDCME13P |
Location: |
Chr.6:149504495-149546102 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIL4 | NM_139126.3 | NP_624311.1 | ||
| AF068298.1 | AAF65187.1 | |||
| AF308286.1 | AAG48254.1 | |||
| AF357880.1 | AAM63961.1 | |||
| AK315388.1 | ||||
| BC016984.1 | AAH16984.1 | |||
| BC020986.1 | AAH20986.1 | |||
| BC064134.1 | AAH64134.1 | |||
| BX537536.1 | CAD97776.1 |
More Information
Set Membership: |
|

Genomic Map