Assay Details
Target Gene Details
Entrez Gene ID: | 147945 |
Gene Name: | NLR family pyrin domain containing 4 |
Gene Aliases: |
CLR19.5, CT58, NALP4, PAN2, PYPAF4, RNH2 |
Location: |
Chr.19:55836578-55881855 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP4 | NM_134444.4 | 5 | 2534 | NP_604393.2 |
| XM_017026344.1 | 4 | 2177 | XP_016881833.1 | |
| XM_017026345.1 | 4 | 2177 | XP_016881834.1 | |
| AF442488.1 | 4 | 2169 | AAL35293.1 | |
| AF479747.1 | 4 | 2181 | AAL87104.1 | |
| AF482706.1 | 4 | 2179 | AAL88672.1 | |
| AK056688.1 | 3 | 2061 | BAB71254.1 | |
| AK315003.1 | 5 | 2534 | ||
| AY072792.1 | 4 | 2172 | AAL68396.1 | |
| BC016443.1 | 3 | 809 | AAH16443.1 | |
| BC050326.1 | 5 | 2208 | AAH50326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644849 | Chr.19:55836319 - 55989037 on Build GRCh38 | Gain |
|
| esv3644851 | Chr.19:55843395 - 56076000 on Build GRCh38 | Gain |
|
| dgv6582n54 | Chr.19:55807428 - 56027874 on Build GRCh38 | Gain |
|
| esv2718994 | Chr.19:55862090 - 55862278 on Build GRCh38 | Deletion |
|
| nsv1064105 | Chr.19:55808050 - 55939166 on Build GRCh38 | Gain |
|
| nsv1057383 | Chr.19:55699665 - 55944662 on Build GRCh38 | Gain |
|
| esv3644852 | Chr.19:55848269 - 55916219 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map