Assay Details
Target Gene Details
Entrez Gene ID: | 2770 |
Gene Name: | G protein subunit alpha i1 |
Gene Aliases: |
Gi |
Location: |
Chr.7:80134824-80219409 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAI1 | NM_002069.5 | 1 | 264 | NP_002060.4 |
| AI878901.1 | 1 | 302 | ||
| AK292953.1 | 1 | 238 | ||
| AL049933.1 | 1 | 242 | CAB43212.2 | |
| BC026326.1 | 1 | 263 | AAH26326.1 |
More Information
Set Membership: |
|

Genomic Map