Assay Details
Target Gene Details
Entrez Gene ID: | 29095 |
Gene Name: | ORMDL sphingolipid biosynthesis regulator 2 |
Gene Aliases: |
HSPC160, MST095, MSTP095, adoplin-2 |
Location: |
Chr.12:55818022-55821175 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORMDL2 | NM_014182.4 | 4 | 813 | NP_054901.1 |
| AB064961.1 | 4 | 767 | BAC11712.1 | |
| AF395705.1 | 3 | 728 | AAM43504.1 | |
| AK223180.1 | 4 | 754 | BAD96900.1 | |
| BC012543.2 | 3 | 723 | AAH12543.1 | |
| BG750383.1 | 2 | 475 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832423 | Chr.12:55687038 - 55859914 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map