Assay Details
Target Gene Details
Entrez Gene ID: | 54557 |
Gene Name: | small glutamine rich tetratricopeptide repeat containing beta |
Gene Aliases: |
SGT2 |
Location: |
Chr.5:65665928-65722821 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGTB | NM_019072.2 | 11 | 1710 | NP_061945.1 |
| XM_005248548.3 | 11 | 1577 | XP_005248605.1 | |
| XM_017009597.1 | 12 | 1846 | XP_016865086.1 | |
| AK096321.1 | 11 | 1665 | BAC04761.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894204 | Chr.5:65586109 - 65714937 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map