Assay Details
Target Gene Details
Entrez Gene ID: | 1054 |
Gene Name: | CCAAT/enhancer binding protein gamma |
Gene Aliases: |
GPE1BP, IG/EBP-1 |
Location: |
Chr.19:33373669-33382686 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPG | NM_001252296.1 | 2 | 1860 | NP_001239225.1 |
| NM_001806.3 | 2 | 1997 | NP_001797.1 | |
| BC013128.1 | 2 | 1915 | AAH13128.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828526 | Chr.19:33368422 - 33387198 on Build GRCh38 | Loss |
|
| esv2758755 | Chr.19:33286168 - 34090656 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map