Assay Details
Target Gene Details
Entrez Gene ID: | 7804 |
Gene Name: | LDL receptor related protein 8 |
Gene Aliases: |
APOER2, HSZ75190, LRP-8, MCI1 |
Location: |
Chr.1:53242364-53328149 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRP8 | NM_001018054.2 | 13 | 2219 | NP_001018064.1 |
| NM_004631.4 | 13 | 2219 | NP_004622.2 | |
| NM_017522.4 | 12 | 1832 | NP_059992.3 | |
| NM_033300.3 | 11 | 1709 | NP_150643.2 | |
| XM_005271173.3 | 14 | 2168 | XP_005271230.1 | |
| XM_005271174.3 | 13 | 1776 | XP_005271231.1 | |
| XM_005271175.3 | 12 | 1737 | XP_005271232.1 | |
| XM_006710881.3 | 14 | 2168 | XP_006710944.1 | |
| XM_006710882.3 | 14 | 2168 | XP_006710945.1 | |
| XM_011542094.2 | 13 | 2042 | XP_011540396.1 | |
| XM_011542095.2 | 12 | 1647 | XP_011540397.1 | |
| XM_011542096.2 | 14 | 2168 | XP_011540398.1 | |
| XM_017002265.1 | 12 | 2003 | XP_016857754.1 | |
| XM_017002266.1 | 14 | 2168 | XP_016857755.1 | |
| XM_017002267.1 | 12 | 1613 | XP_016857756.1 | |
| XM_017002268.1 | 13 | 1652 | XP_016857757.1 | |
| AK096482.1 | 11 | 1487 | ||
| AK122887.1 | 5 | 2679 | ||
| BC051836.1 | 13 | 2124 | AAH51836.2 | |
| CD300289.1 | 3 | 332 | ||
| D50678.1 | 13 | 2084 | BAA09328.1 | |
| Z75190.1 | 12 | 1592 | CAA99509.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011750 | Chr.1:53226981 - 53261502 on Build GRCh38 | Gain |
|
| nsv829871 | Chr.1:53200871 - 53383048 on Build GRCh38 | Loss |
|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
| nsv509258 | Chr.1:53259839 - 53285955 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map