Assay Details
Target Gene Details
Entrez Gene ID: | 9683 |
Gene Name: | NEDD4 binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.16:48538726-48610214 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N4BP1 | NM_153029.3 | 7 | 2960 | NP_694574.3 |
| XM_011523482.1 | 6 | 2857 | XP_011521784.1 | |
| AB014515.1 | 7 | 2960 | ||
| AK001464.1 | 6 | 1370 | ||
| AK026937.1 | 6 | 2051 | ||
| BC108288.1 | 6 | 2521 | AAI08289.1 | |
| BC152472.1 | 7 | 2960 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2976n100 | Chr.16:48497260 - 48544190 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map