Assay Details
Target Gene Details
Entrez Gene ID: | 85476 |
Gene Name: | G elongation factor mitochondrial 1 |
Gene Aliases: |
COXPD1, EFG, EFG1, EFGM, EGF1, GFM, hEFG1 |
Location: |
Chr.3:158644497-158692572 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFM1 | NM_001308164.1 | NP_001295093.1 | ||
| NM_001308166.1 | NP_001295095.1 | |||
| NM_024996.5 | NP_079272.4 | |||
| XM_006713795.1 | XP_006713858.1 | |||
| AF309777.1 | AAK58877.1 | |||
| AF367998.1 | AAK53402.1 | |||
| AK000780.1 | ||||
| AK092293.1 | ||||
| AK315031.1 | ||||
| BC049210.1 | AAH49210.1 | |||
| KU178816.1 | ||||
| KU178817.1 |
Target Gene Details
Entrez Gene ID: | 56925 |
Gene Name: | latexin |
Gene Aliases: |
ECI, TCI |
Location: |
Chr.3:158666414-158672722 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LXN | NM_020169.3 | 1 | 72 | NP_064554.3 |
| XM_017006876.1 | 1 | 101 | XP_016862365.1 | |
| AF282626.1 | AAF82807.1 | |||
| BC005346.1 | 1 | 72 | AAH05346.1 | |
| BC008438.1 | 1 | 54 | AAH08438.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763312 | Chr.3:158475723 - 158702799 on Build GRCh38 | Gain |
|
| nsv1014255 | Chr.3:158641071 - 159050753 on Build GRCh38 | Gain |
|
| nsv1004124 | Chr.3:158428926 - 159090849 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map