Assay Details
Target Gene Details
Entrez Gene ID: | 2977 |
Gene Name: | guanylate cyclase 1 soluble subunit alpha 2 |
Gene Aliases: |
GC-SA2, GUC1A2 |
Location: |
Chr.11:106674012-107018445 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCY1A2 | NM_000855.2 | 7 | 2281 | NP_000846.1 |
| NM_001256424.1 | 8 | 2374 | NP_001243353.1 | |
| BC130484.1 | 7 | 1987 | ||
| BC130488.1 | 7 | 1987 | ||
| BC144033.1 | 8 | 2050 | ||
| X63282.1 | 7 | 2281 | CAA44921.1 | |
| Z50053.1 | 8 | 2374 | CAA90393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556229 | Chr.11:106696291 - 106711896 on Build GRCh38 | Loss |
|
| nsv1052674 | Chr.11:104111970 - 107043401 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map