Assay Details
Target Gene Details
Entrez Gene ID: | 6945 |
Gene Name: | MLX, MAX dimerization protein |
Gene Aliases: |
MAD7, MXD7, TCFL4, bHLHd13 |
Location: |
Chr.17:42567060-42573203 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLX | NM_170607.2 | 8 | 1269 | NP_733752.1 |
| NM_198204.1 | 8 | 1107 | NP_937847.1 | |
| NM_198205.1 | 7 | 1017 | NP_937848.1 | |
| XM_017024991.1 | 7 | 1156 | XP_016880480.1 | |
| AF203978.1 | 8 | 1057 | AAF14638.1 | |
| AF213666.1 | 7 | 1005 | AAG40145.1 | |
| AF213667.1 | 8 | 1095 | AAG40146.1 | |
| AF213668.1 | 8 | 1257 | AAG40147.1 | |
| AK000150.1 | 8 | 1066 | BAA90977.1 | |
| AK290258.1 | ||||
| AK303221.1 | 8 | 990 | ||
| BC010689.2 | 8 | 1092 | AAH10689.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map