Assay Details
Target Gene Details
Entrez Gene ID: | 8034 |
Gene Name: | solute carrier family 25 member 16 |
Gene Aliases: |
D10S105E, GDA, GDC, HGT.1, ML7, hML7 |
Location: |
Chr.10:68479961-68527523 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A16 | NM_001324312.1 | 9 | 1446 | NP_001311241.1 |
| NM_001324314.1 | 10 | 1600 | NP_001311243.1 | |
| NM_001324315.1 | 9 | 2275 | NP_001311244.1 | |
| NM_001324317.1 | 8 | 1284 | NP_001311246.1 | |
| NM_152707.3 | 9 | 1418 | NP_689920.1 | |
| NR_136737.1 | 8 | 2153 | ||
| XM_011540211.2 | XP_011538513.1 | |||
| AK123772.1 | 8 | 2154 | ||
| AK298510.1 | 10 | 1493 | ||
| BC030266.1 | 9 | 1369 | AAH30266.1 | |
| M31659.1 | 10 | 1437 | AAA36329.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762871 | Chr.10:68441880 - 68510208 on Build GRCh38 | Gain |
|
| dgv920n100 | Chr.10:68441868 - 68493125 on Build GRCh38 | Gain |
|
| dgv919n100 | Chr.10:68400084 - 68752800 on Build GRCh38 | Gain |
|
| nsv1044477 | Chr.10:68428357 - 68628622 on Build GRCh38 | Gain |
|
| nsv551434 | Chr.10:68384017 - 68503822 on Build GRCh38 | Loss |
|
| nsv1050943 | Chr.10:68414663 - 68511675 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map