Assay Details
Target Gene Details
Entrez Gene ID: | 11051 |
Gene Name: | nudix hydrolase 21 |
Gene Aliases: |
CFIM25, CPSF5 |
Location: |
Chr.16:56429136-56451349 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT21 | NM_007006.2 | 7 | 1743 | NP_008937.1 |
| BX537360.1 | 7 | 1747 | CAD97606.1 | |
| BX647665.1 | 3 | 1363 | CAI46057.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833242 | Chr.16:56259354 - 56446840 on Build GRCh38 | Loss |
|
| nsv833243 | Chr.16:56375371 - 56549461 on Build GRCh38 | Gain |
|
| esv3638695 | Chr.16:56413924 - 56440936 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map