Assay Details
Target Gene Details
Entrez Gene ID: | 6158 |
Gene Name: | ribosomal protein L28 |
Gene Aliases: |
L28 |
Location: |
Chr.19:55385932-55392085 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL28 | NM_000991.4 | 5 | 4046 | NP_000982.2 |
| NM_001136134.1 | 4 | 4240 | NP_001129606.1 | |
| NM_001136135.1 | 5 | 735 | NP_001129607.1 | |
| AB209627.1 | 2 | 5459 | BAD92864.1 | |
| AK025866.1 | 1 | 1842 | ||
| BC011582.1 | 4 | 4025 | AAH11582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953624 | Chr.19:55302033 - 55625034 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map