Assay Details
Target Gene Details
Entrez Gene ID: | 11277 |
Gene Name: | three prime repair exonuclease 1 |
Gene Aliases: |
AGS1, CRV, DRN3, HERNS |
Location: |
Chr.3:48465520-48467645 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TREX1 | NM_007248.3 | 1 | 438 | NP_009179.2 |
| NM_016381.5 | 1 | 438 | NP_057465.1 | |
| NM_033629.4 | 1 | 438 | NP_338599.1 | |
| AF151105.3 | 1 | 95 | AAD48774.2 | |
| AJ243797.1 | CAB50866.1 | |||
| AL137745.1 | 1 | 438 | ||
| BQ933154.1 | 1 | 92 | ||
| DA364183.1 | 1 | 128 | ||
| DA850044.1 | 1 | 123 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834684 | Chr.3:48344209 - 48551890 on Build GRCh38 | Gain |
|
| dgv8315n54 | Chr.3:48464432 - 48478412 on Build GRCh38 | Loss |
|
| nsv590242 | Chr.3:48392157 - 48518906 on Build GRCh38 | Loss |
|
| nsv508917 | Chr.3:48400224 - 48499357 on Build GRCh38 | Insertion |
|

Genomic Map