Assay Details
Target Gene Details
Entrez Gene ID: | 121227 |
Gene Name: | leucine rich repeats and immunoglobulin like domains 3 |
Gene Aliases: |
LIG3 |
Location: |
Chr.12:58869351-58920540 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRIG3 | NM_001136051.2 | 19 | 3120 | NP_001129523.1 |
| NM_153377.4 | 19 | 3539 | NP_700356.2 | |
| XM_011537893.2 | 19 | 3211 | XP_011536195.1 | |
| AK074921.1 | 10 | 2039 | BAC11295.1 | |
| AK130083.1 | 5 | 1007 | ||
| AK292857.1 | 19 | 3520 | ||
| AL833710.1 | 8 | 1876 | ||
| AY358288.1 | 19 | 3482 | AAQ88655.1 | |
| AY358295.1 | 19 | 3120 | AAQ88662.1 | |
| AY505340.1 | 19 | 3290 | AAR98629.1 | |
| BC126169.1 | 19 | 3435 | ||
| BC126171.1 | 19 | 3435 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759902 | Chr.12:58628947 - 58929264 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map