Assay Details
Target Gene Details
Entrez Gene ID: | 57649 |
Gene Name: | PHD finger protein 12 |
Gene Aliases: |
PF1 |
Location: |
Chr.17:28905250-28951490 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHF12 | NM_001033561.1 | 15 | 3334 | NP_001028733.1 |
| XM_005258015.3 | 14 | 2623 | XP_005258072.1 | |
| XM_005258016.3 | 14 | 2643 | XP_005258073.1 | |
| XM_011525078.2 | 14 | 2935 | XP_011523380.1 | |
| XM_011525079.2 | 14 | 3218 | XP_011523381.1 | |
| XM_017024903.1 | 14 | 3382 | XP_016880392.1 | |
| XM_017024904.1 | 14 | 2902 | XP_016880393.1 | |
| AB040956.1 | 10 | 1855 | BAA96047.1 | |
| AK024290.1 | 8 | 1660 | BAB14875.1 | |
| AK091441.1 | 1 | 2897 | ||
| AK291420.1 | 15 | 2806 | ||
| AK294056.1 | 14 | 2763 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457719 | Chr.17:28867616 - 29014758 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map