Assay Details
Target Gene Details
Entrez Gene ID: | 80725 |
Gene Name: | SRC kinase signaling inhibitor 1 |
Gene Aliases: |
P140, SNIP |
Location: |
Chr.17:38530015-38607166 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRCIN1 | NM_025248.2 | 19 | 4691 | NP_079524.2 |
| XM_017025169.1 | 23 | 5783 | XP_016880658.1 | |
| XM_017025170.1 | 21 | 4981 | XP_016880659.1 | |
| XM_017025171.1 | 20 | 4858 | XP_016880660.1 | |
| XM_017025172.1 | 19 | 4756 | XP_016880661.1 | |
| AB051471.1 | 13 | 3203 | BAB21775.1 | |
| AB127405.1 | BAD03968.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428341 | Chr.17:37855347 - 38600766 on Build GRCh38 | Gain+Loss |
|
| esv27695 | Chr.17:38460087 - 38532791 on Build GRCh38 | Gain |
|
| nsv953893 | Chr.17:38527166 - 38590447 on Build GRCh38 | Deletion |
|
| nsv469701 | Chr.17:38394019 - 38545635 on Build GRCh38 | Loss |
|
| esv2758688 | Chr.17:37706608 - 38600766 on Build GRCh38 | Gain+Loss |
|
| nsv833436 | Chr.17:38455845 - 38669136 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map